Thermo Scientific 75003662 Fixed-Angle Rotor; 4x250 mL
Representative image only.
Thermo Scientific Mfr # 75003662 – Item # EW-17708-30
$6,772.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)10000
  • Max RCF x g15316
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted250 mL
  • Number Of Tubes Accepted4
  • Max Tube Length (mm)125
  • Max Tube Diameter (mm)62
  • DescriptionFixed-Angle Rotor; 4x250 mL
More About this Item
Choose the rotor compatible with your Thermo Scientific centrifuge and application needs.
Increase performance with versatility, speed and a robust, corrosion-free carbon fiber design by using the Thermo Scientific™ Fiberlite™ F14-6 x 250LE Fixed Angle Rotor, which supports a range of capacities and applications.
Compatible with: Thermo Scientific™ Sorvall™ Legend™ XT/XF, Heraeus™ Multifuge™ X3/X3F and SL 40F centrifuges.