Thermo Scientific 8162SC pH electrode, Refillable/DJ/Glass; Screw Cap
Representative image only.
Thermo Scientific Mfr # 8162SC – Item # EW-05719-01
Drift less than 0.002 pH per day
PRODUCT OPTIONS
$471.60USD / Each
$524.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsGeneral purpose-Tris buffers
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionSleeve
  • Reference ElectrolyteROSS
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ConnectorScrew cap
  • DescriptionpH electrode, Refillable/DJ/Glass; Screw Cap
Includes
50-mL of 3 M KCl filling solution and 1-m cable. Order additional fill solution separately.
More About this Item
These provide fast, accurate, and stable pH readings. Ideal for food, pharmaceuticals, or biological samples. Platinum wire prevents contamination with silver and mercury ions—excellent for use in tris buffer. Double ceramic junction design protects the inner reference solution and prolongs the life of the electrode. Readings stabilize to 0.01 pH in less than 30 seconds—even with a 50degC temperature change. Electrodes read pH from 0 to 14. Max temperature is 90degC for glass electrodes and 80degC for epoxy. All electrodes have BNC connectors.
Models 05718-20 and 05721-26 have a pin-tip connector, which is identical to the pin-tip portion of a U.S. Standard connector.