Thermo Scientific 88880001 RT2 Basic Digital Hot Plate, Ceramic Top; 230V

Representative image only.
Thermo Scientific Mfr # 88880001 – Item # EW-86576-02
Safely and accurately heat your samples
  • Sleek low-profile design saves valuable bench space
  • Ceramic-coated aluminum top provides excellent temperature uniformity and is corrosion resistant
  • Digital control with PID microprocessor
  • "Hot top" warning visually alerts lab personnel of potential hazard even when unit is shut off
  • Built-in overtemperature shuts off if top plate temperature reaches 842°F (450°C)
  • Three user-selectable temperature control modes: optimal, fast, and slow
  • Display allows you to simultaneously view set point and actual temperature
  • Optional nonslip heating bath and transparent safety shield (sold separately)
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Top Plate Diameter (in)5 1/2
  • Top Plate Diameter (cm)13.97
  • Top Plate MaterialCeramic-coated Aluminum
  • Min Temperature (° F)75
  • Max Temperature (° F)662
  • Min Temperature (° C)20
  • Max Temperature (° C)350
  • Max Load Capacity (lbs)55
  • Max Load Capacity (kg)25
  • Length (in)11 2/5
  • Width (in)6 19/64
  • Height (in)3 29/32
  • Length (cm)28.956
  • Width (cm)16.002
  • Height (cm)9.906
  • Power (VAC)230
  • Power (Hz)50/60
  • Power (watts)600
  • DescriptionRT2 Basic Digital Hot Plate, Ceramic Top; 230V
  • Warranty2 year
Two year warrantyConformite Europeenne

Key Features

  • Sleek low-profile design saves valuable bench space
  • Ceramic-coated aluminum top provides excellent temperature uniformity and is corrosion resistant
  • Digital control with PID microprocessor
  • "Hot top" warning visually alerts lab personnel of potential hazard even when unit is shut off
  • Built-in overtemperature shuts off if top plate temperature reaches 842°F (450°C)
  • Three user-selectable temperature control modes: optimal, fast, and slow
  • Display allows you to simultaneously view set point and actual temperature
  • Optional nonslip heating bath and transparent safety shield (sold separately)
  • 2 year warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat