Thermo Scientific 900012 Filling Solution, sodium, 5/cs
Representative image only.
Thermo Scientific Mfr # 900012 – Item # EW-05730-50
Filling solutions for use with pH electrodes and reference electrodes
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
PRODUCT OPTIONS
$213.75USD / Case of 5
Include Accessories Today!
SPECS & DESCRIPTION
Specifications & Description
  • Ion-Selective ElectrodeSodium
  • DescriptionFilling Solution, sodium, 5/cs
Key Features
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
More About this Item
All filling solutions for reference electrodes come in cartons of one 60-mL or five 50-mL bottles.