Thermo Scientific 910686 pH Buffer Solution, 6.86, Clear; 475 mL Bottle
Representative image only.
Thermo Scientific Mfr # 910686 – Item # EW-55350-56
Common calibration values are olor-coded for ease of use
  • Choose from pint-size or portable 60-mL bottles
  • Traceable to NIST standards
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Value6.86
  • Buffer Kit Values6.86
  • Volume (mL)475
  • ColorClear
  • DescriptionpH Buffer Solution, 6.86, Clear; 475 mL Bottle
Key Features
  • Choose from pint-size or portable 60-mL bottles
  • Traceable to NIST standards
More About this Item
Thermo Scientific Orion pH buffers are available in 475-mL (pint) bottles and in portable 5 x 60 mL convenience packs. Portable pH buffers are easier to carry than larger bottles, minimize wasted buffer, and provide more volume than pouches.
Each buffer is color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.