Thermo Scientific Epredia™ High-Quality Dehydrant, Reagent Alcohol Blend, 100%, 1 Gallon; 4/Cs

Representative image only.
Thermo Scientific Mfr # 9111 – Item # EW-48801-10
Designed for tissue processing and staining of histological and cytological specimens
  • Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants
  • Blend of 90% Ethyl alcohol, 4.5% Methyl alcohol, 5.5% Isopropyl alcohol
  • 1 month warranty

PRODUCT OPTIONS

$176.00USD / Case of 4

SPECS & DESCRIPTION

Specifications & Description

  • Concentration100%
  • Element/IonReagent Grade Alcohol Blend
  • FormLiquid
  • SynonymsDenatured Ethyl alcohol
  • Volume (Gal)4
  • PackagingEpredia™ HDPE Bottles
  • DOT InformationEthanol Solution; Hazard Class 3, Packing Group II
  • CAS Number67-63-0 (Isopropyl alcohol) 64-17-5 (Ethyl alcohol) 67-56-1 (Methyl alcohol)
  • UN NumberU1170
  • Boiling Point C (° C)78.5
  • Melting Point C (° C)-114.1
  • DescriptionDehydrant, Reagent Alcohol Blend, 100%, 1 Gallon; 4/Cs
  • Warranty1 month
1 Month Warranty

Key Features

  • Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants
  • Blend of 90% Ethyl alcohol, 4.5% Methyl alcohol, 5.5% Isopropyl alcohol
  • 1 month warranty

More About this Item

Choose the dehydrating solvent that works best with your staining procedure. Packaged in easy-to-pour high-density polyethylene containers, Epredia™ High-Quality Dehydrants are safer and lighter than glass bottles.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat