Thermo Scientific Formaldehyde Solution, 37%, 4 x 1 Gal

Representative image only.
Thermo Scientific Mfr # 9311 – Item # EW-48800-00
Easy-to-pour container minimizes splashing and reduces spills
  • Fix and store tissue specimens in histological applications
  • 1 month warranty

$258.50USD / Case of 4

SPECS & DESCRIPTION

Specifications & Description

  • Concentration0.37
  • FormLiquid
  • Volume (Gal)4
  • PackagingPoly bottle
  • CAS Number50-00-0
  • DescriptionFormaldehyde Solution, 37%, 4 x 1 Gal
  • Warranty1 month
1 Month Warranty

Key Features

  • Fix and store tissue specimens in histological applications
  • 1 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat