Thermo Scientific A2-24D Replacement Comb for Electrophoresis Gels, 20 cm W, 24 Wells; 1/EA
Representative image only.
Thermo Scientific Mfr # A2-24D – Item # EW-28553-32
SPECS & DESCRIPTION
Specifications & Description
  • Comb Thickness (mm)1.5
  • Number of Wells24
  • DescriptionReplacement Comb for 28553-20,-30 24 wells 1.5mm gel
More About this Item
Use combs to produce the wells you need for your electrophoresis application. Select the comb based on the gel width and the number of wells.