Thermo Scientific A2-BP Owl Recirculating Cell, 20 x 25 cm gel size

Representative image only.
Thermo Scientific Mfr # A2-BP – Item # EW-28553-30
Longer path length for more detailed DNA/RNA analysis
  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs

PRODUCT OPTIONS

$1,174.00USD / Each
Spectrum Chemical Agarose
Spectrum Chemical Agarose
$126.00 - $2,554.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Solution Required (mL)2300
  • Gel Length (cm)25
  • Gel Width (cm)20
  • Gel Thickness (mm)1.5
  • Cell Width (in)11
  • Cell Height (in)4
  • Cell Depth (in)14.75
  • Cell Width (cm)27.94
  • Cell Height (cm)10.16
  • Cell Depth (cm)37.47
  • DescriptionOwl Recirculating Cell, 20 x 25 cm gel size

Key Features

  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs

More About this Item

Self-recirculating models feature barbed fitting ports for buffer exchange via an external recirculating pump to minimize buffer breakdown and maintain constant pH.


What's included: buffer chamber, UV transparent gel tray with gasket end gates, safety lid with attached power leads, and built-in leveling base. All combs are 1.5-mm thick.
Item 28553-30 also includes one each 16-, 24-, and 36-well combs.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat