Thermo Scientific Barnstead 06.5040 Smart2Pure Water Reservior Tank with Level Control; 30 Liter
Representative image only.
Thermo Scientific Barnstead Mfr # 06.5040 – Item # EW-99299-11
Optimize performance of your Thermo Scientific Barnstead Water Purification System
  • Compatible with the Thermo Scientific Barnstead Water Purification System
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)30
  • MaterialPolyethylene (PE)
  • Compatible WithSmart2Pure 12 Water Purification Systems
  • Height (in)23.5
  • Diameter (in)15
  • DescriptionSmart2Pure Water Reservior Tank with Level Control; 30 Liter
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Compatible with the Thermo Scientific Barnstead Water Purification System
  • 1 year warranty
More About this Item
Choose from a wide range of accessories and replacement parts for your water purification system.
These opaque polyethylene reservoirs feature a large opening and conical bottom outlet for complete draining and easy cleaning/disinfection. Recirculating pump protects water from bacterial growth and maintains low conductivity values. Tank requires overflow and filter for use, please see accessories. Wall-mounting bracket is optional to free up precious lab space.