Thermo Scientific Barnstead D440046 Single-cartridge deionizer for
Representative image only.
Thermo Scientific Barnstead Mfr # D440046 – Item # EW-99292-40
Reduce the level of scaling ions entering your still—clean your still less often!
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)10.5
  • Depth (in)25.5
  • Width (in)7.5
  • DescriptionSingle-cartridge deionizer for
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Deionizers provide a pretreated water supply to any Mega-Pure still sold above. Both feature a temperature-compensated purity meter that continuously monitors water purity and indicates when cartridges are exhausted; a flow control valve for precise output flow control; and a built-in drain valve for fast, no-spill cartridge replacement. Systems withstand pressure from 10 to 100 psi.
Systems include 5 ft of 1¼2" ID vinyl drain tubing; 5 ft of 7¼16" ID vinyl output tubing; 3¼8" OD inlet connection; and a 7-ft cord with three-prong plug for 120 VAC operation. Order optional solenoid valve 99292-50 to provide your system with an automatic shutoff when output water resistivity falls below 50 kΩ-cm. Order a still adapter kit to install systems directly to your still.
Single-cartridge Mega-Pure deionizer economically removes dissolved inorganic matter from most water supplies. Measures 101¼2"W x 251¼2"H x71¼2"D. Use with high-capacity cartridge only (sold separately).