Thermo Scientific Barnstead D4801 Type I Water Cartridge Kit
Representative image only.
Thermo Scientific Barnstead Mfr # D4801 – Item # EW-99258-00
Keep your system operating precisely
  • Use with quad-style Nanopure® systems
  • Kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagent grade water
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionType I Water Cartridge Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • Use with quad-style Nanopure® systems
  • Kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagent grade water
  • 1 year warranty
More About this Item
Kit contains one 01505-22, one 01505-24, and two 01505-29.