Thermo Scientific Barnstead D5028 Type I cartridge kit, four module
Representative image only.
Thermo Scientific Barnstead Mfr # D5028 – Item # EW-99258-21
Select pretreatment, deionization, or organic-free cartridges, depending upon
your application
  • For use with E-Pure systems
  • Produce MΩ-cm resistivity, Type I reagentgrade water
  • Four module
PRODUCT OPTIONS
$1,045.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionType I cartridge kit, four module
Includes
One pretreatment cartridge (01505-22), one deionization cartridge (01505-24), and two high purity deionization cartridges (01505-29).
Key Features
  • For use with E-Pure systems
  • Produce MΩ-cm resistivity, Type I reagentgrade water
  • Four module
More About this Item
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
The kit includes all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water.