Thermo Scientific Barnstead D7421 EASYpure Compact Reverse Osmosis System, 120 VAC
Representative image only.
Thermo Scientific Barnstead Mfr # D7421 – Item # EW-99271-00
Small footprint – easily mount on the wall or use on the benchtop! Provides flow rate of 10 liters per hour.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (L/hour)10
  • Inlet Port3/8\" quick disconnect (included)
  • Max Inlet Water Temperature (° C)50
  • DescriptionEASYpure Compact Reverse Osmosis System, 120 VAC
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These systems are ideal for pretreating your EASYpure® deionization systems. They remove up to 95% of ionized impurities and 99% of bacteria and organics with molecular weight greater than 300 dalton. Includes a 6.5-L reservoir with air filter to prevent contamination from airborne impurities during storage.
Use these automated systems to simplify operation, monitor membrane performance, and prolong the life of the thin film membrane. In addition, an automatic flush cleans the thin film membrane to eliminate build-up.
System includes: 6.5-L reservoir, 6-ft cord and plug, and 3/8" quick-disconnect fitting. A 30-L reservoir is available, order separately. To wall mount the system, order mounting bracket 99261-58.