Thermo Scientific Barnstead E3452 Pura-Lite E3454 Resistivity Indicators
Representative image only.
Mfr # E3452 – Item # EW-01504-38
A low-cost alternative to digital purity meters
  • Designed for use with B-Pure systems
  • 1 year warranty
PRODUCT OPTIONS
$300.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Inlet Port1/2\" NPT(F)
  • Resistivity Threshold1 MΩ
  • DescriptionPura-Lite E3454 Resistivity Indicators
  • Warranty1 year
1 Year Warranty
Key Features
  • Designed for use with B-Pure systems
  • 1 year warranty
More About this Item
These indicators can be mounted directly on top of single- or doublecartridge housings with 1⁄2" NPT(F) connections.
Green light indicates water resistivity is above the threshold; red light warns if resistivity falls below threshold. Operates on 120 VAC power.