Thermo Scientific Barnstead FL703X2 Final Filter for 99254-02 Dispenser
Representative image only.
Thermo Scientific Barnstead Mfr # FL703X2 – Item # EW-99261-60
Use with Nanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
SPECS & DESCRIPTION
Specifications & Description
  • Compatible withEasypure II, Nanopure Infinity, replacement filter for Remote Dispenser 99254-02
  • Max Flow Rate (LPM)1.8
  • DescriptionFinal Filter for 99254-02 Dispenser
More About this Item
Final filters remove particulates down to 0.2 µm to provide bacteria- free water. Constructed of hollow fiber material, filters do not contain any wetting agents or surfactants. Filters are radiation sterilized and individually sealed.
Autoclavable.