Thermo Scientific Barnstead NANOpure Water Purification System, Analytical
Representative image only.
Thermo Scientific Barnstead Mfr # D11901 – Item # EW-99277-00
  • Automatic calibration verification and optional NIST traceable calibration module, 99275-56, lets you calibrate the system in the field
  • Purity up to 18.2 MΩ-cm — exceeds ASTM, CAP, and NCCLS Type 1 requirements
  • Ensure the organic carbon content of your water meets your most stringent standards with TOC monitoring models
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Inlet Port3/8\" OD compression
  • Max Inlet Water Temperature (° F)120
  • Min Inlet Water Temperature (° F)40
  • Max Inlet Water Temperature (° C)49
  • Min Inlet Water Temperature (° C)4
  • Max Inlet Pressure (PSI)100
  • DescriptionNANOpure Water Purification System, Analytical
  • Warranty1 year
1 Year Warranty
Key Features
  • Automatic calibration verification and optional NIST traceable calibration module, 99275-56, lets you calibrate the system in the field
  • Purity up to 18.2 MΩ-cm — exceeds ASTM, CAP, and NCCLS Type 1 requirements
  • Ensure the organic carbon content of your water meets your most stringent standards with TOC monitoring models
  • 1 year warranty
More About this Item
Thermo Scientific Barnstead NANOpure™ reagent grade water purification systems are designed to support tissue culture, PCR, electrophoresis, atomic absorption (AA), ICP, ICP/MS, ion chromatography (IC), gas chromatography (GC), and HPLC. Standby mode recirculates water for 10 minutes every hour during inactive periods.
Additional standard features include compensated and uncompensated purity measurement, volumetric or timed dispensing of ultrapure water, automatic sanitization cycle, 0.2 µm absolute final filtration, and integral pressure regulating valve.
NANOpure Analytical. The perfect system for your most sensitive analytical procedures including AA, ICP, ICP/MS, IC, GC, and high-wavelength HPLC. Produces up to 1.5 liters of ultrapure water per minute.