Thermo Scientific BioBasic 73205-101030 SCX Column - Length 100mm, 1.0mm ID, 5µm particle size
Representative image only.
Thermo Scientific BioBasic Mfr # 73205-101030 – Item # EW-01901-81
Stable polymeric coating suitable for separation and retention of basic and other cationic species
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSCX
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)1
  • DescriptionSCX Column - Length 100mm, 1.0mm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic SCX Columns utilize a polymeric sulfonic acid ion exchange ligand that is bound to a highly pure, base-deactivated silica that creates a stable polymeric coating, even in demanding pH conditions (pH range of 2 to 8). Proven in the separation of cationic proteins and peptides, as well as neurotransmitters, vitamins, and quaternary amine herbicides. These columns are extremely reliable, providing reproducible results run after run.