Thermo Scientific BioBasic 73205-150065 SCX KAPPA Capillary Column - Length 150mm, 75µm ID, 5µm particle size
Representative image only.
Thermo Scientific BioBasic Mfr # 73205-150065 – Item # EW-01901-92
Provide high efficiency and sensitivity – ideal for LC/MS analysis
  • KAPPA Capillary Columns are especially suitable for small sample, proteomic separations
  • Nanobore Columns provide higher sensitivity for nanospray applications
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSCX
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)0.075
  • DescriptionSCX KAPPA Capillary Column - Length 150mm, 75µm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • KAPPA Capillary Columns are especially suitable for small sample, proteomic separations
  • Nanobore Columns provide higher sensitivity for nanospray applications
  • 1 month warranty
More About this Item
BioBasic KAPPA Capillary Columns are manufactured to meet the demanding sensitivities of high performance proteomics and LC/MS analyses. Small internal diameters (75µm to 500µm) provide lower flow rates, required by electrospray ionization methods. These capillaries are able to detect, identify, and quantify analytes in the picomole and femtomole levels. Manufactured from inert tubing and unions compatible wuth 10 - 32 male nuts and 1/16" OD capillary tubing. Columns also have a stainless steel sleeve for added strength and ease of handling.