Thermo Scientific BioBasic 73305-307846 SEC 60 Column - 300mm by 7.8mm, 60 Angstrom pore size
Representative image only.
Thermo Scientific BioBasic Mfr # 73305-307846 – Item # EW-01902-09
Provide superior separation of water soluble compounds
  • Ideally suited for sample clean-up
  • Analyte separation over a wide range of molecular weights
  • Long, efficient column life
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSilica
  • DescriptionSEC 60 Column - 300mm by 7.8mm, 60 Angstrom pore size
  • Warranty1 month
1 Month Warranty
Key Features
  • Ideally suited for sample clean-up
  • Analyte separation over a wide range of molecular weights
  • Long, efficient column life
  • 1 month warranty
More About this Item
BioBasic SEC (Size Exclusion Chromatography) columns are made with a mechanically rigid silica that does not swell or shrink with changes in solvent, providing higher efficiencies than polymer-based columns. The highly base-deactivated 5µm silica is polymer-coated to ensure separation only occurs on the basis of molecular size. Columns provide highly efficient separation of compounds with a molecular weight of 100 to 10,000,000.
Each lot of silica used in the production of BioBasic SEC columns is tested with a range of proteins to confirm accuracy. Each column is also tested for compliance with efficiency specifications and is shipped with a test certificate with both the column and silica efficiency data.
BioBasic SEC columns are ideal for gel filtration separation of proteins an