Thermo Scientific CF-1 NanoDrop™ Spectrophotometer Calibration Fluid, 0.5 mL Ampule
Representative image only.
Thermo Scientific Mfr # CHEM-CF-1 – Item # EW-83060-90
Optimize your NanoDrop with these calibration accessories
  • Ensure accuracy of your spectrophotometer with these calibration accessories
PRODUCT OPTIONS
$110.00USD / Each
$120.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)0.5
  • MaterialAqueous Potassium Dichromate Solution
  • Compatible WithNanoDrop 8000, NanoDrop Lite, NanoDrop 2000, and NanoDrop 2000c spectrophotometers
  • DescriptionCF-1 NanoDrop™ Spectrophotometer Calibration Fluid, 0.5 mL Ampule
Key Features
  • Ensure accuracy of your spectrophotometer with these calibration accessories
More About this Item
Choose from a wide range of calibration accessories to keep your Thermo Scientific™ NanoDrop™ spectrophotometers and fluorometers running accurately and efficiently. Spectrophotometers sold separately.
This aqueous potassium dichromate solution is perfect for calibration of Thermo Scientific NanoDrop™ spectrophotometers. Solution is roughly ten times more concentrated than other potassium dichromate solutions used for calibration checks of spectrophotometers. Intended for single use, solution is packaged in a 0.5 ml snap-top glass ampule.
Available for sale and use in the United States only.