Thermo Scientific CF-8 NanoDrop™ Spectrophotometer Calibration Check Kit
Representative image only.
Thermo Scientific Mfr # CHEM-CF-8 – Item # EW-83060-92
Optimize your NanoDrop with these calibration accessories
  • Ensure accuracy of your spectrophotometer with these calibration accessories
PRODUCT OPTIONS
$169.65USD / Each
$188.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)2 x 0.5
  • MaterialAqueous Potassium Dichromate Solution
  • Compatible WithNanoDrop 8000 Spectrophotometer
  • DescriptionCF-8 NanoDrop™ Spectrophotometer Calibration Check Kit
Includes
two CF-1 ampules and an 8-Well PCR tube strip.
Key Features
  • Ensure accuracy of your spectrophotometer with these calibration accessories
More About this Item
Choose from a wide range of calibration accessories to keep your Thermo Scientific™ NanoDrop™ spectrophotometers and fluorometers running accurately and efficiently. Spectrophotometers sold separately.
This calibration kit contains everything you need to confirm your calibration of the Thermo Scientific NanoDrop™ 8000 Spectrophotometer. Includes potassium dichromate solution and one 8-well PCR tube strip.
Available for sale and use in the United States only.