Thermo Scientific CIC0001448 External Temperature Probe, Stainless Steel, PT1000, 0-400°C
Representative image only.
Thermo Scientific Mfr # CIC0001448 – Item # EW-04600-92
Enhance performance and safety of your stirrers and hotplates
  • Optimize use of your hotplates and stirrers
  • Ideal for use with Cimarec and Supernuova hotplates and hotplate stirrers
  • 3 year warranty
PRODUCT OPTIONS
$107.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)32
  • Max Temperature (° F)752
  • Min Temperature (° C)0
  • Max Temperature (° C)400
  • DescriptionExternal Probe Stainless Steel, PT1000
  • Warranty3 year
3 Year Warranty
Key Features
  • Optimize use of your hotplates and stirrers
  • Ideal for use with Cimarec and Supernuova hotplates and hotplate stirrers
  • 3 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories suitable for use with the Thermo Scientific™ Cimarec+™ and Super-Nuova+™ stirrers, hotplates, or hotplate stirrers. Choose accessories compatible with your equipment.
External temperature probe works with both Cimarec+ and SuperNuova+ hot plates, stirrers, and stirring hot plates. Standard stainless steel is suitable for most laboratory environments. Maximum temperature is 752°F (400°C).