This item is now discontinued. Please browse similar items

Thermo Scientific Cimarec 50094711 i Mono Direct Stirrer 230V Euro

Representative image only.
Thermo Scientific Cimarec Mfr # 50094711 – Item # EW-04656-25
Memory function stores last speed settings
  • Electromagnetic drive means no moving parts and no maintenance

SPECS & DESCRIPTION

Specifications & Description

  • Top Plate Length (in)5
  • Top Plate Width (in)5
  • Top Plate Length (cm)12.7
  • Top Plate Width (cm)12.7
  • Top Plate MaterialStainless Steel
  • Low Speed (rpm)130
  • High Speed (rpm)990
  • No. Of Stirring Positions1
  • Max Stirring Volume (Liters)3
  • Length (in)7 13/16
  • Width (in)6
  • Height (in)1
  • Length (cm)19.85
  • Width (cm)15.24
  • Height (cm)4.13
  • Plug TypeEuropean
  • Power (VAC)230
  • Power (Hz)50/60
  • Descriptioni Mono Direct Stirrer 230V Euro

Includes

6-ft cord with a power supply.

Key Features

  • Electromagnetic drive means no moving parts and no maintenance

More About this Item

Use these Thermo Scientific Cimarec® stirrers for growing microorganisms, dissolving nutrient mediums and solids, and titration applications.

Thermo Scientific Cimarec® Stirrers accommodate a variety of vessel shapes including Erlenmeyer flasks and beakers. Stirring speed remains constant even if viscosity of the medium changes. The powder-coated stainless steel housing protects unit from spills and dirt.

Note: Not for use in hazardous areas; not immersible.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat