Thermo Scientific Finnpipette 4651020 Pipette , Fixed at 10 uL; Each
Representative image only.
Thermo Scientific Finnpipette Mfr # 46510200N – Item # EW-25014-54
A new level of sophistication created to meet the demands of modern laboratories
  • The first pipette to have an antimicrobial surface treatment to prevent contamination
  • Adjustable finger rest for a customized feel based on your preferences
  • Large, easy-to-read digital display on adjustable-volume models
  • Comfortable, lightweight design with smooth pipetting action
  • Soft-tip ejection
PRODUCT OPTIONS
$403.00USD / Each
$70.00 - $81.00USD / Each
for an additional
$70.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)10
  • Max Volume (mL)0.01
  • Max Dispensing (mL)0.01
  • Accuracy (%)±0.90%
  • Reproducibility±0.80%
  • Number Of Channels1
  • DescriptionPipette , Fixed at 10 uL; Each
Key Features
  • The first pipette to have an antimicrobial surface treatment to prevent contamination
  • Adjustable finger rest for a customized feel based on your preferences
  • Large, easy-to-read digital display on adjustable-volume models
  • Comfortable, lightweight design with smooth pipetting action
  • Soft-tip ejection
  • Color-coded plunger for quick and easy identification
  • 5-year warranty with web registration