Thermo Scientific Haake 399-0200 High-range portable viscometer, range 30 to 400,000 cP

Representative image only.
Thermo Scientific Haake Mfr # 399-0200 – Item # EW-08705-02
Small, battery-operated design for quick field tests and comparative measurements
  • Rotational viscometers provide quick, exact, and reliable measurements for a variety of fluid types ranging from paints and shampoos to sauces and oils
  • Simple, one button operation eliminates set-up time and allows hassle-free testing
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min Viscosity Range (cP)30
  • Max Viscosity Range (cP)400000
  • Reproducibility±1% full scale
  • Viscosity Accuracy±5% full-scale
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Sample Volume Needed (mL)150
  • Rpm65.5
  • BatteryFour AA batteries
  • DescriptionHigh-range portable viscometer, range 30 to 400,000 cP
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Rotational viscometers provide quick, exact, and reliable measurements for a variety of fluid types ranging from paints and shampoos to sauces and oils
  • Simple, one button operation eliminates set-up time and allows hassle-free testing
  • 1 year warranty

More About this Item

These portable digital viscometers are lightweight and can be operated handheld or on a stand (order separately) for your convenience. Converting the handheld to use on a stand is only a matter of seconds - simply place in the special instrument holder. Use for quick viscosity tests for process optimization, machine adjustment, or to ensure your product is within specification.
By displaying viscosity on a digital display, human error associated with manual timing and misreading analog dials are eliminated. Possible handling errors as well as service information are also shown on the display.
Pushing a button to turn the unit on; push the button again to select the rotor type, then begin measuring. A rotor rotating at a constant speed is immersed in the fluid to be tested; the fluid's resistance to the rotation is correlated to measuring the fluid viscosity. Three different-sized rotors are included with the viscometer for multiple application capabilities.

What's inlcuded: three rotors, carrying case, and four AA batteries. Model 08705-02 also includes one 150-mL sample cup.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat