Thermo Scientific 75005703 Round Bucket Inserts, Tall, 90°, Rma x 165 mm for Swinging Bucket Rotor 17701-30
Representative image only.
Thermo Scientific Mfr # 75005703 – Item # EW-17701-31
Exceptional capacity and flexible processing adds value to any lab
PRODUCT OPTIONS
$2,740.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Capacity (mL)50
  • Tube Capacity8 x 50 mL
  • DescriptionRound Bucket Inserts, Tall, 90°, Rmax 165mm for Swinging Bucket Rotor 17701-30
More About this Item
Make the most of your centrifuge with buckets, lids, and inserts.