Thermo Scientific Heraeus 75005721 Centrifuge Tube Adapter, Sealed Bucket for M10; 2/Pk
Representative image only.
Thermo Scientific Heraeus Mfr # 75005721 – Item # EW-17701-42
Exceptional capacity and flexible processing adds value to any lab
  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe
PRODUCT OPTIONS
$1,606.00USD / Each
Often Bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Tube Capacity4 x standard microplates or -2 x deepwell microplates
  • Max Tube Length (mm)33
  • DescriptionCentrifugeTubeAdapter,SealedBucketforM10;2/Pk
Key Features
  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe
More About this Item
Conforming to the latest safety standards, the Megafuge 8 is a compact bench centrifuge that satisfies many sample running options. The secure push-button Auto-Lock® rotor exchange system with glove-friendly design allows you to change out rotors with ease, requiring only one hand! The SMARTSpin imbalance detection system optimizes acceleration, braking, and load imbalance for smooth runs and reproducible separations.
Centrifuge features one-touch operation and pre-saved programming with password protection. The control panel with highly visible backlit display is glove- and detergent-friendly. Brushless motor provides quiet performance and a stress free work environment.