Thermo Scientific Heraeus 75007210 Small Benchtop Centrifuge; 120 VAC

Representative image only.
Thermo Scientific Heraeus Mfr # 75007210 – Item # EW-17701-20
Exceptional capacity and flexible processing adds value to any lab
  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe

$3,302.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Speed (rpm)15000
  • Min Speed (rpm)300
  • Max RCF x g21693
  • Rotors IncludedNo
  • RefrigeratedNo
  • Timer Range1 minute to 9 hours, 99 minutes
  • Max Capacity (mL)580
  • Tube Capacity4 x 145 mL (adapters available for other volumes)-2 x 2 standard microplates-2 x 1 depwell microplates
  • Power (VAC)120
  • Power (Hz)50/60
  • DescriptionSmall Benchtop Centrifuge; 120 VAC

Key Features

  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe

More About this Item

Conforming to the latest safety standards, the Megafuge 8 is a compact bench centrifuge that satisfies many sample running options. The secure push-button Auto-Lock® rotor exchange system with glove-friendly design allows you to change out rotors with ease, requiring only one hand! Choose from several rotor types, including: the Fiberlite carbon rotor— with a fifteen year warranty it is one of the most robust rotor options available; the swinging bucket rotor for microplates or buckets; or the standard fixed angle rotor which holds up to 600-mL capacities. The rotors with ClickSeal™ (sealed) lids provide ultimate chemical resistance and biocontainment protection while eliminating the need for screw caps and clips. The SMARTSpin imbalance detection system optimizes acceleration, braking, and load imbalance for smooth runs and reproducible separations.

Centrifuge features one-touch operation and pre-saved programming with password protection. The control panel with highly visible backlit display is glove- and detergent-friendly. Brushless motor provides quiet performance and a stress free work environment.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat