Thermo Scientific Heraeus 75007555 Swinging bucket, use with rotor 17702-60
Representative image only.
Thermo Scientific Heraeus Mfr # 75007555 – Item # EW-17702-61
  • A round bucket for 4 x 100 mL max
  • Adapters for all types of tubes such as DIN, blood sample, disposable conical and micro tubes from 1.5 to 100 mL
  • An aerosol tight cap is available as an option sold separately
$461.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSwinging bucket, use with rotor 17702-60
Key Features
  • A round bucket for 4 x 100 mL max
  • Adapters for all types of tubes such as DIN, blood sample, disposable conical and micro tubes from 1.5 to 100 mL
  • An aerosol tight cap is available as an option sold separately
More About this Item
This swinging bucket rotor is compatible with Thermo Scientific™ Sorvall™ Primo™ and Heraeus™ Primo™ Centrifuges. This rotor has a max speed of 4,000 rpm, max RCF of 2,585x g, and a max radius of 14.5 cm.