Thermo Scientific Heraeus M10 (75005706) Sealed Microplate Bucket
Representative image only.
Thermo Scientific Heraeus Mfr # 75005721 – Item # EW-17701-27
Exceptional capacity and flexible processing adds value to any lab
  • Use with Thermo Scientific™ Sorvall™ ST 8, Heraeus™ Megafuge™ 8 and SL 8 centrifuges
  • K Factor = 13,103
  • Radius: 119 mm
PRODUCT OPTIONS
$1,632.00USD / Each
Often Bought with...
Cole-Parmer ThinTouch™ Nitrile Gloves
$9.00 - $79.50USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)4400
  • Max RCF x g2576
  • Tube Capacity4 x standard microplates or -2 x deepwell microplates
  • DescriptionSealed Microplate Bucket
Key Features
  • Use with Thermo Scientific™ Sorvall™ ST 8, Heraeus™ Megafuge™ 8 and SL 8 centrifuges
  • K Factor = 13,103
  • Radius: 119 mm
More About this Item
Make the most of your centrifuge with buckets, lids, and inserts.