Thermo Scientific Heraeus TX-200 Swinging Bucket Rotor; 4 x 180 mL
Representative image only.
Thermo Scientific Heraeus Mfr # 75003658 – Item # EW-17709-10
  • Rotor "bar" only; buckets and sealing lids not included
PRODUCT OPTIONS
$1,444.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)5500
  • Max RCF x g5580
  • Rotor TypeSwing-out
  • Tube Sizes Accepted180 mL
  • Number Of Tubes Accepted4
  • Max Tube Length (mm)123
  • Max Tube Diameter (mm)56.5
  • DescriptionSwingingBucketRotor;4x180mL
Key Features
  • Rotor "bar" only; buckets and sealing lids not included
More About this Item
Choose the rotor compatible with your Thermo Scientific centrifuge and application needs.
Accommodate a wide range of processing volumes with the Thermo Scientific™ TX-200 4 x 180mL Swinging Bucket Rotor, Buckets, and Sealing Lids (rotor, buckets, and lids sold separately).
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Heraeus™ Multifuge™ X1, Heraeus Megafuge™ 16 Centrifuge Series