Thermo Scientific Hypersil 28205-014001 BDS C8 Drop-In Guard Cartridge - Length 10mm, 4.0mm / 4.6mm ID, 5µm particle size; Pack of 4
Representative image only.
Thermo Scientific Hypersil Mfr # 28205-014001 – Item # EW-01885-98
Protect and prolong the life of your column
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Column Particle Size (µm)5
  • DescriptionBDS C8 Drop-In Guard Cartridge - Length 10mm, 4.0mm / 4.6mm ID, 5µm particle size; Pack of 4
  • Warranty1 month
1 Month Warranty
Key Features
  • Convenient and economical
  • Available in a variety of stationary phases and particle sizes
  • 1 month warranty
More About this Item
These drop-in guard cartridges provide effective protection, prolonging the life of your analytical columns. Available in a wide variety of phases in a 10mm design with minimal impact on retention time. Useful for light to moderately contaminated samples, these cartridges should be replaced after contamination is detected. Sold in packs of 4.