Thermo Scientific Hypersil 30503-253030 SAS (C1) Column - Length 250mm, 3.0mm ID, 3µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 30503-253030 – Item # EW-01888-54
High-efficiency columns with proven results and reproducibility
  • Suitable for SAS (C1) phase
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSAS (C1)
  • Column Particle Size (µm)3
  • Tubing Inside Diameter (ID) (mm)3
  • DescriptionSAS (C1) Column - Length 250mm, 3.0mm ID, 3µm particle size
  • Warranty1 month
1 Month WarrantyISO9001 Certified Supplier
Key Features
  • Suitable for SAS (C1) phase
  • 1 month warranty
More About this Item
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil SAS (C1) Columns are the least retentive of the Hypersil columns and are useful for ion-pair separations. Exhibiting good stability, these columns show selectivity for polar analytes and multifunctional compounds. Stable in pH range of 2 to 8.