Thermo Scientific Hypersil 30505-012101 SAS Drop-In Guard Cartridges, 2.1mm ID, 5um; 4/Pk
Representative image only.
Thermo Scientific Hypersil Mfr # 30505-012101 – Item # EW-01888-57
Protect and prolong the life of your column
  • Easily trap contaminants such as solvent and sample debris
  • Convenient and economical
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSAS (C1)
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionSAS Drop-In Guard Cartridges, 2.1mm ID, 5um; 4/Pk
  • Warranty1 month
1 Month WarrantyISO9001 Certified Supplier
Key Features
  • Easily trap contaminants such as solvent and sample debris
  • Convenient and economical
  • 1 month warranty
More About this Item
Drop-In Guard Cartridges will extend the life and performance of your column. These cartridges provide an economical and efficient way of trapping contaminants such as solvent and sample debris. Drop-in design makes for easy guard and column replacement — simply position between the injector and analytical column to remove damaging components before they reach the column. The 10-mm length offers maximum protection with minimal retention.