Thermo Scientific Hypersil 30803-153030 CPS (Cyano) Column - Length 150mm, 3.0mm ID, 3µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 30803-153030 – Item # EW-01889-22
High-efficiency columns with proven results and reproducibility
  • Suitable for Cyano phase
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseCyano
  • Column Particle Size (µm)3
  • Tubing Inside Diameter (ID) (mm)3
  • DescriptionCPS (Cyano) Column - Length 150mm, 3.0mm ID, 3µm particle size
  • Warranty1 month
1 Month WarrantyISO9001 Certified Supplier
Key Features
  • Suitable for Cyano phase
  • 1 month warranty
More About this Item
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil CPS and CPS-2 (Cyano) Columns provide a more rapid mobile phase equilibration compared to silica. These columns are one of the least retentive reversed-phase materials and are useful in the separation of polar compounds. Hypersil CPS-2 is endcapped. Stable in pH range of 2 to 8.