Thermo Scientific Hypersil 34105-102130 SAX Column - Length 100mm, 2.1mm ID, 5µm particle size
Representative image only.
Thermo Scientific Hypersil Mfr # 34105-102130 – Item # EW-01890-96
High-efficiency columns with proven results and reproducibility
  • Suitable for SAX Quaternary Amine phase
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseSAX
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionSAX Column - Length 100mm, 2.1mm ID, 5µm particle size
  • Warranty1 month
1 Month WarrantyISO9001 Certified Supplier
Key Features
  • Suitable for SAX Quaternary Amine phase
  • 1 month warranty
More About this Item
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil SAX Quaternary Amine Columns are highly stable silica-based strong anion exchange columns — ideally suited for the analysis of smaller organic molecules such as nucleotides and organic acids. Stable in pH range of 2 to 9.