Thermo Scientific M-20 Microplate Swinging Rotor; 6 standard or 2 deepwell
Representative image only.
Thermo Scientific Mfr # 75003624 – Item # EW-17709-12
  • Includes standard open carriers
PRODUCT OPTIONS
$2,190.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)4000
  • Max RCF x g2272
  • Rotor TypeSwing-out
  • Tube Sizes Accepted2 x 3 (6) standard microplates or 2 x 1 (2) deepwell plates
  • Number Of Tubes Accepted6
  • DescriptionMicroplateSwingingRotor;6standardor2deepwell
Key Features
  • Includes standard open carriers
More About this Item
Choose the rotor compatible with your Thermo Scientific centrifuge and application needs.
Capacity for 2 x 3 (6) standard microplates or 2 x 1 (2) deepwell plates.