Thermo Scientific MicroClick 30 x 2 Fixed-Angle Microtube Rotor; 30 x 2mL
Representative image only.
Thermo Scientific Mfr # 75005719 – Item # EW-17701-45
  • ClickSeal™ biocontainment lid provides ultimate chemical resistance and protection while eliminating the need for screw caps and clips
PRODUCT OPTIONS
$3,096.00USD / Each
Often Bought with...
Cole-Parmer ThinTouch™ Nitrile Gloves
$9.00 - $67.50USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)14000
  • Max RCF x g21693
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted1.5 or 2.0 mL microtubes
  • Number Of Tubes Accepted30
  • Description30x2Fixed-AngleMicrotubeRotor;30x2mL
Key Features
  • ClickSeal™ biocontainment lid provides ultimate chemical resistance and protection while eliminating the need for screw caps and clips
More About this Item
Choose the rotor compatible with your Thermo Scientific centrifuge and application needs.
Perform micro-volume separations for applications such as nucleic acid preparation at up to 14,000 rpm/21,694 x g with the Thermo Scientific™ MicroClick 30 x 2 Fixed Angle Microtube Rotor.
Compatible with Heraeus™ Megafuge™ 8, Sorvall™ ST8, and SL8 centrifuges.