Thermo Scientific Nalgene 2135-5303 Top Works 53B Cap, 1/4" ID (2), 1/8" ID (1)
Representative image only.
Thermo Scientific Nalgene Mfr # 2135-5303 – Item # EW-13070-08
  • Top Works™ closure systems and NALGENE® bottles and carboys create the ideal aseptic fluid transfer system!
PRODUCT OPTIONS
$433.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size53B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID6.4 mm
  • DescriptionTop Works 53B Cap, 1/4" ID (2), 1/8" ID (1)
Key Features
  • Top Works™ closure systems and NALGENE® bottles and carboys create the ideal aseptic fluid transfer system!
More About this Item
Flexible Top Works™ Closures are available in solid, two-port and three-port versions. Closures are polypropylene (PP). Platinum-cured silicone tubing is fused to a silicone stopper insert which forms a one-piece closure system. Autoclavable and leakproof.
Call Us At1-847-549-7600
Live chat