Thermo Scientific Nalgene 2624-0020 Stainless Steel Dolly
Representative image only.
Thermo Scientific Nalgene Mfr # 2624-0020 – Item # EW-50301-95
Makes moving small tanks and carboys easy
  • Constructed of durable stainless steel
  • Noncorrosive and resistant to acids and bases
  • Casters do not leave marks on floor
  • Autoclavable
$905.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Compatible WithThermo Scientific Nalhene Bioprocess Tanks
  • Height (in)6.5
  • Diameter (in)20.5
  • Height (cm)16.5
  • Diameter (cm)52.1
  • DescriptionSS dolly, 20-1/2"ID x 6-1/2"H
Key Features
  • Constructed of durable stainless steel
  • Noncorrosive and resistant to acids and bases
  • Casters do not leave marks on floor
  • Autoclavable
More About this Item
Stainless steel dolly helps transport tanks up to 115 liters. Compatible with tanks 06318-70 and 50301-52, tanks sold separately. Measures 20 1⁄2" ID x 6 1⁄2"H (52.1 x 16.5 cm)