Thermo Scientific Nalgene 3118-0085 Round-bottom tubes; 85 mL; PC; 10/pack

Representative image only.
Thermo Scientific Nalgene Mfr # 3118-0085 – Item # EW-06328-70
Separate materials without leaks
  • Ideal for high-speed centrifugation qnd lipis extraction

PRODUCT OPTIONS

$192.00USD / Pkg of 10

BUY MORE & SAVE

  • 1-5$192.00USD / Pkg of 10
  • 6-29$172.80USD / Pkg of 10
  • 30-119$163.20USD / Pkg of 10
  • 120+$153.60USD / Pkg of 10

SPECS & DESCRIPTION

Specifications & Description

  • Max RCF x g50000
  • AutoclavableYes
  • Tube Capacity (mL)85
  • Tube MaterialPolycarbonate
  • Tube ColorClear
  • Tube Bottom ShapeRound
  • Tube Length (mm)105.7
  • Tube Outside Diameter (OD) (mm)38.2
  • Cap IncludedYes
  • Cap StyleSealing Cap
  • Cap MaterialPolypropylene
  • Closure Size Number33
  • GraduationsNone
  • DescriptionRound-bottom tubes; 85 mL; PC; 10/pack

Key Features

  • Ideal for high-speed centrifugation qnd lipis extraction

More About this Item

FEP tubes are ideal for high-speed centrifugation of aggressive chemicals. Sealing caps ensure leakproof performance at speeds greater than 10,000 RCF x g and are recommended when spinning hazardous materials. Tubes are autoclavable adn withstand temperatures from -100 to 150°C (-148 to 302°F).

Note: FEP tubes must be filled to 100% of capacity and centrifuges under refrigeration; do not use at room temperature.

Thermo Scientific Nalgene® Nalge Thermo Scientific Nunc International
PTFE, TEFZEL® E.I. DuPont de Nemours & Co.
VITON® Dupont Dow Elastomers L.L.C.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat