Thermo Scientific Nalgene 3120-1000 Centrifuge bottle; 1000 mL; PPCO; 4/pack

Representative image only.
Thermo Scientific Nalgene Mfr # 3120-1000 – Item # EW-06105-60

$222.00USD / Pkg of 4

SPECS & DESCRIPTION

Specifications & Description

  • Max RCF x g7100
  • AutoclavableYes
  • Tube Capacity (mL)100
  • Tube MaterialPolypropylene
  • Tube ColorNatural
  • Tube Bottom ShapeRound
  • Tube Length (mm)184.5
  • Tube Outside Diameter (OD) (mm)97.5
  • Cap IncludedYes
  • Cap MaterialPolypropylene
  • Closure Size Number63
  • DescriptionCentrifuge bottle; 1000 mL; PPCO; 4/pack

More About this Item

Choose from PPCO or PC. Bottles are injection blown—no “pinchmarks” that could contribute to failure. To use bottles at maximum speeds without leakage or to spin hazardous materials, order sealing caps under accessories. Models 06106-40 and 06105-60 fit the following rotors: Beckman JS-5.2, JS-4.2, JS-4.0, and JS-3.0 (when used with Beckman 344040 adapter). All models fit Sorvall HG-4L, H-4000, H-6000A (no adapter needed) and IEC 266, 949, and 966 (when used with appropriate IEC adapter). Thermo Scientific Nalgene®.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat