Thermo Scientific Nalgene 5306-0130 polycarbonate vacuum plate for 06761-25, -30
Representative image only.
Thermo Scientific Nalgene Mfr # 5306-0130 – Item # EW-06763-02
  • 1 year warranty
$186.00USD / Each
BUY MORE & SAVE
  • 1-5$186.00USD / Each
  • 6-29$167.40USD / Each
  • 30-119$158.10USD / Each
  • 120+$148.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Overall Height (in)1
  • Overall Height (cm)2.5
  • Outside Diameter (OD) (in)14 1/4
  • Outside Diameter (OD) (cm)36.2
  • Descriptionpolycarbonate vacuum plate for 06761-25, -30
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use with jars 06761-12, -25, and -30, or with bell jars that have an OD up to 12''. Plates include 1/4'' tubing connector and neoprene gasket. Maximum vacuum rating is 29''Hg. NALGENE.