Thermo Scientific Nalgene 6149-0001 barbed bulkhead fitting kit, 1/2" tubing ID
Representative image only.
Thermo Scientific Nalgene Mfr # 6149-0001 – Item # EW-06259-00
$41.00USD / Pkg of 2
BUY MORE & SAVE
  • 1-5$41.00USD / Pkg of 2
  • 6-29$36.90USD / Pkg of 2
  • 30-119$34.85USD / Pkg of 2
  • 120+$32.80USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • AutoclavableYes
  • Tubing ID12.7 mm
  • Descriptionbarbed bulkhead fitting kit, 1/2" tubing ID
More About this Item
Fitting Kits include two PP fittings, two acetal nuts, two silicone gaskets, thermoplastic elastomer (TPE) port cap, instructions, and template. Autoclavable.