Thermo Scientific Nalgene 6370-0005 Biohazard Waste Container, 5 gal.
Representative image only.
Thermo Scientific Nalgene Mfr # 6370-0005 – Item # EW-81401-05
A convenient way to dispose of infectious waste
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Absorption5 gal./ 22\" x 30\" bag
  • OD (in)11
  • Height (in)15
  • Compliance29 CFR Part 1910.1030.
  • DescriptionBiohazard Waste Container, 5 gal.
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These leakproof plastic containers feature a port opening for one-handed operation — limits exposure to biohazard/infectious waste, including bloodborne pathogens. The container is reusable, autoclavable, and won’t rust, dent, or puncture. Made of PPCO (polypropylene copolymer). Biohazard liner bags sold separately right. Approval: OSHA Standard 29 CFR Part 1910.1030