Thermo Scientific Nalgene PC Storage CryoBox with Vials Bundle, 81 x 1-2 mL Vials; 4/Pk
Representative image only.
Thermo Scientific Nalgene Mfr # 5026-0909 – Item # EW-06754-54
  • CryoBox holds 81 vials of 1-2 mL
PRODUCT OPTIONS
$188.00USD / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held81
  • Tube Size1.0 to 2.0 mL
  • Lowest Set Temperature (° C)-196
  • ColorBlue and Clear
  • Height (in)2
  • Height (cm)5.1
  • DescriptionPC Storage CryoBox Bundle, 81 x 1-2 mL Vials; 4/Pk
Includes
Cryogenic Vials Storage Box and 50 Cryogenic Vials
Key Features
  • CryoBox holds 81 vials of 1-2 mL
More About this Item
Nalgene cryogenic vial storage boxes are ideal for mechanical freezers or liquid nitrogen. Polycarbonate (PC) boxes are vented, allowing rapid cooling of contents. Gridded insert and special lid with one-way fit make sample inventory foolproof. Boxes withstand temperatures from –196 to 121°C. Autoclavable.