This item is now discontinued. We recommend the following as suggested replacements: EW-62300-16
Thermo Scientific Nalgene RTU Safety Wash Bottles 500 mL Ethanol LDPE; 24/Cs
Representative image only.
Thermo Scientific Nalgene Mfr # 2428-0502 – Item # EW-42011-02
Protect all workers with global safety guidelines
  • Nozzles deliver straight stream for precision dispensing
  • One-piece molded closure and stem eliminate leaks
  • Self-venting feature eliminates dripping and sputtering
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)500
  • MaterialLow Density Polyethyene (LDPE)
  • Cap ColorWhite
  • Label TypeEthanol
  • DescriptionSafety Wash Bottles 500 mL Ethanol LDPE; 24/Cs
Key Features
  • Nozzles deliver straight stream for precision dispensing
  • One-piece molded closure and stem eliminate leaks
  • Self-venting feature eliminates dripping and sputtering
More About this Item
Prevent chemical accidents in the lab with properly labeled containers. The chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications is clearly printed on each “Right-to-Understand” (RTU) wash bottle.
LDPE Wash Bottles have color-coded closures for quick identification of chemical contents. Compatible with methanol, isopropanol, ethanol, distilled water, sodium hypochlorite, and deionized water.