Thermo Scientific Nalgene RTU Safety Wash Bottles 500 mL n-Hexane FEP; 4/Cs
Representative image only.
Thermo Scientific Nalgene Mfr # 2429-0503 – Item # EW-42011-13
Protect all workers with global safety guidelines
  • Nozzles deliver straight stream for precision dispensing
  • One-piece molded closure and stem eliminate leaks
  • Self-venting feature eliminates dripping and sputtering
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)500
  • MaterialFluorinated Ethylene Propylene (FEP)
  • Cap ColorNatural
  • Label Typen-Hexane
  • DescriptionSafety Wash Bottles 500 mL n-Hexane FEP; 4/Cs
Key Features
  • Nozzles deliver straight stream for precision dispensing
  • One-piece molded closure and stem eliminate leaks
  • Self-venting feature eliminates dripping and sputtering
More About this Item
Prevent chemical accidents in the lab with properly labeled containers. The chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications is clearly printed on each “Right-to-Understand” (RTU) wash bottle.
FEP Wash Bottles feature permanently etched GHS chemical safety information. Can be used for methyl ethyl ketone, n-hexane, toluene, xylene, methylene chloride, and acetone. Autoclavable.