Thermo Scientific Nalgene Wash Bottle, FEP, 125 mL, 1/pk

Representative image only.
Thermo Scientific Nalgene Item # EW-06052-20
Won't contaminate reagents
  • Resistant to most chemicals
  • One-piece closure with spout prevents leaks
  • Autoclavable

PRODUCT OPTIONS

$181.00USD / Each

BUY MORE & SAVE

  • 1-4$181.00USD / Each
  • 5-9$162.90USD / Each
  • 10+$144.80USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)125
  • Capacity (oz)4
  • MaterialFEP
  • Cap MaterialEthylene Tetrafluoroethylene (ETFE)
  • Cap / Stopper Size24 mm
  • Cap ColorNatural
  • Bottle ColorTransparent
  • AutoclavableYes
  • Outside Diameter (OD) (in)2
  • Outside Diameter (OD) (cm)5.1
  • Overall Height (in)6 1/4
  • Overall Height (cm)15.9
  • DescriptionWash Bottle, FEP, 125 mL, 1/pk

Key Features

  • Resistant to most chemicals
  • One-piece closure with spout prevents leaks
  • Autoclavable

More About this Item

FEP bottles resist most chemicals and will not contaminate reagents—use to dispense strong solvents. A one-piece closure with spout prevents leaks.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat